COMH4211763F Combi-Blocks, Inc.

2-Chloro-1-(1,2,3,4,5,6,7,8-octahydro-phenanthren-9-yl)-ethanone

  • MDL number MFCD13705054
  • Cas Number : 861329-27-3
  • Molecular Weight : 262.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6680
Availability details for
Quantity
  • Availability
  • Add to Cart