COMH42117640 Combi-Blocks, Inc.

tert-Butyl [(4-hydroxy-3,5-dimethylpyridin-2-yl)methyl]carbamate

  • MDL number MFCD31557141
  • Cas Number : 2200383-59-9
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6681
Availability details for
Quantity
  • Availability
  • Add to Cart