COMH42117649 Combi-Blocks, Inc.

2-[(5-Chloroquinolin-8-yl)oxy]ethanamine dihydrochloride

  • MDL number MFCD31583455
  • Cas Number : 2270905-27-4
  • Molecular Weight : 295.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((5-chloroquinolin-8-yl)oxy)ethan-1-amine dihydrochloride
Inchi Code 1S/C11H11ClN2O.2ClH/c12-9-3-4-10(15-7-5-13)11-8(9)2-1-6-14-11;;/h1-4,6H,5,7,13H2;2*1H
Inchi Key HSDVDUOPFFSRLX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6737
Availability details for
Quantity
  • Availability
  • Add to Cart