COMH4211764C Combi-Blocks, Inc.

1-(2-Chloro-6-fluorobenzyl)-1h-indole-2,3-dione

  • MDL number MFCD01248191
  • Cas Number : 314025-87-1
  • Molecular Weight : 289.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6748
Availability details for
Quantity
  • Availability
  • Add to Cart