COMH4211764F Combi-Blocks, Inc.

2-(1H-Pyrazol-1-yl)quinoline-4-carboxylic acid

  • MDL number MFCD11544195
  • Cas Number : 1152718-85-8
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-pyrazol-1-yl)-4-quinolinecarboxylic acid
Inchi Code 1S/C13H9N3O2/c17-13(18)10-8-12(16-7-3-6-14-16)15-11-5-2-1-4-9(10)11/h1-8H,(H,17,18)
Inchi Key FKVCCXZRAYMRMV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6766
Availability details for
Quantity
  • Availability
  • Add to Cart