COMH42117651 Combi-Blocks, Inc.

Rhodamine b isothiocyanate

  • MDL number MFCD00041787
  • Cas Number : 36877-69-7
  • Molecular Weight : 536.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6808
Availability details for
Quantity
  • Availability
  • Add to Cart