COMH42117652 Combi-Blocks, Inc.

(5-Hydroxy-benzofuran-3-yl)-pyridin-2-yl-methanone

  • MDL number MFCD23134769
  • Cas Number : 953423-73-9
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6816
Availability details for
Quantity
  • Availability
  • Add to Cart