COMH4211765F Combi-Blocks, Inc.

3-Bromo-1-(2,4-difluorophenyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD31581345
  • Cas Number : 2270905-26-3
  • Molecular Weight : 288.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6889
Availability details for
Quantity
  • Availability
  • Add to Cart