COMH42117663 Combi-Blocks, Inc.

N-[2-(4-Aminophenoxy)ethyl]acetamide

  • MDL number MFCD11891572
  • Cas Number : 22404-15-5
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[2-(4-aminophenoxy)ethyl]acetamide
Inchi Code 1S/C10H14N2O2/c1-8(13)12-6-7-14-10-4-2-9(11)3-5-10/h2-5H,6-7,11H2,1H3,(H,12,13)
Inchi Key WOVQEYXDOCRQID-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6913
Availability details for
Quantity
  • Availability
  • Add to Cart