COMH42117674 Combi-Blocks, Inc.

[(3,5-Dimethylisoxazol-4-yl)methyl]dimethylamine hydrochloride

  • MDL number MFCD31557146
  • Molecular Weight : 190.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6995

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6995
Availability details for
Quantity
  • Availability
  • Add to Cart