COMH42117679 Combi-Blocks, Inc.

2-(Aminomethyl)-3,5-dimethyl-1-(pyridin-3-ylmethyl)pyridin-4(1h)-one dihydrochloride

  • MDL number MFCD31557142
  • Cas Number : 2270906-70-0
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7024
Availability details for
Quantity
  • Availability
  • Add to Cart