COMH4211767D Combi-Blocks, Inc.

6-(9,10-Dimethoxy-5,11-dioxoisoindolo[2,1-a]quinazolin-6(5h,6ah,11h)-yl)hexanoic acid

  • MDL number MFCD28142498
  • Cas Number : 1630763-97-1
  • Molecular Weight : 424.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7062
Availability details for
Quantity
  • Availability
  • Add to Cart