COMH4211767E Combi-Blocks, Inc.

4-(1-Propen-2-yl)biphenyl

  • MDL number MFCD20384577
  • Cas Number : 34352-84-6
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7065
Availability details for
Quantity
  • Availability
  • Add to Cart