COMH42117684 Combi-Blocks, Inc.

Ethyl 3-methyl-5-oxo-1-phenyl-4,5-dihydro-1h-pyrazole-4-carboxylate

  • MDL number MFCD00194465
  • Cas Number : 29711-06-6
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-methyl-5-oxo-1-phenyl-4,5-dihydro-1H-pyrazole-4-carboxylate
Inchi Code 1S/C13H14N2O3/c1-3-18-13(17)11-9(2)14-15(12(11)16)10-7-5-4-6-8-10/h4-8,14H,3H2,1-2H3
Inchi Key PFJJREYHEKKQLY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7100
Availability details for
Quantity
  • Availability
  • Add to Cart