COMH42117693 Combi-Blocks, Inc.

1-tert-Butyl 4-ethyl 2-oxopiperidine-1,4-dicarboxylate

  • MDL number MFCD29054241
  • Cas Number : 1313498-26-8
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7164
Availability details for
Quantity
  • Availability
  • Add to Cart