COMH42117694 Combi-Blocks, Inc.

Methyl 2-cyano(2,2-dimethyltetrahydro-4h-pyran-4-ylidene)acetate

  • MDL number MFCD30238685
  • Cas Number : 14389-99-2
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7171
Availability details for
Quantity
  • Availability
  • Add to Cart