COMH421176A0 Combi-Blocks, Inc.

Dspe-peg4-dbco

  • MDL number MFCD30828684
  • Cas Number : 2112738-14-2
  • Molecular Weight : 1282.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C71H116N3O15P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-41-70(78)86-60-65(89-71(79)42-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)61-88-90(80,81)87-52-49-73-68(76)47-51-83-54-56-85-58-57-84-55-53-82-50-46-67(75)72-48-45-69(77)74-59-64-39-34-33-37-62(64)43-44-63-38-35-36-40-66(63)74/h33-40,65H,3-32,41-42,45-61H2,1-2H3,(H,72,75)(H,73,76)(H,80,81)/t65-/m1/s1
Inchi Key RIQFURRJENJBHO-IDKUVDLHSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7237
Availability details for
Quantity
  • Availability
  • Add to Cart