COMH421176A3 Combi-Blocks, Inc.

N-(4-Aminophenyl)isonicotinamide

  • MDL number MFCD02585680
  • Cas Number : 13116-08-0
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7248
Availability details for
Quantity
  • Availability
  • Add to Cart