COMH421176A5 Combi-Blocks, Inc.

3-(Ethylsulfinyl)-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31583440
  • Cas Number : 1707737-39-0
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(ethylsulfinyl)-1H-1,2,4-triazol-5-amine
Inchi Code 1S/C4H8N4OS/c1-2-10(9)4-6-3(5)7-8-4/h2H2,1H3,(H3,5,6,7,8)
Inchi Key CMFVNJFQAMZSLW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7269
Availability details for
Quantity
  • Availability
  • Add to Cart