COMH421176A7 Combi-Blocks, Inc.

Active-mono-sulfone-peg8-cid

  • MDL number MFCD30458030
  • Cas Number : 2055048-45-6
  • Molecular Weight : 767.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7278
Availability details for
Quantity
  • Availability
  • Add to Cart