COMH421176AB Combi-Blocks, Inc.

3-Dimethylaminomethyl-4-hydroxy-benzoic acid

  • MDL number MFCD20641579
  • Cas Number : 103095-57-4
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7296
Availability details for
Quantity
  • Availability
  • Add to Cart