COMH421176B8 Combi-Blocks, Inc.

2-Oxo-5-phenyl-2,3-dihydro-1h-imidazole-4-carbaldehyde

  • MDL number MFCD28248782
  • Cas Number : 1820638-81-0
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-5-phenyl-1H-imidazole-4-carbaldehyde
Inchi Code 1S/C10H8N2O2/c13-6-8-9(12-10(14)11-8)7-4-2-1-3-5-7/h1-6H,(H2,11,12,14)
Inchi Key QKOWJQXUBBJHPU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7336
Availability details for
Quantity
  • Availability
  • Add to Cart