COMH421176BA Combi-Blocks, Inc.

(2E)-3-(1-Methyl-1h-indol-5-yl)acrylic acid

  • MDL number MFCD16618455
  • Cas Number : 1449582-78-8
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-(1-methyl-1H-indol-5-yl)-2-propenoic acid
Inchi Code 1S/C12H11NO2/c1-13-7-6-10-8-9(2-4-11(10)13)3-5-12(14)15/h2-8H,1H3,(H,14,15)/b5-3+
Inchi Key IZZVPUQKWOCNSS-HWKANZROSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7338

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7338
Availability details for
Quantity
  • Availability
  • Add to Cart