COMH421176C0 Combi-Blocks, Inc.

6-(3-Fluorophenyl)-3-methylimidazo[2,1-b][1,3]thiazole-2-carboxylic acid hydrochloride

  • MDL number MFCD22123344
  • Cas Number : 1131580-14-7
  • Molecular Weight : 312.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7367
Availability details for
Quantity
  • Availability
  • Add to Cart