COMH421176C3 Combi-Blocks, Inc.

Ethyl (2e)-3-[(4-methoxybenzyl)amino]but-2-enoate

  • MDL number MFCD31581330
  • Cas Number : 2088565-26-6
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7394
Availability details for
Quantity
  • Availability
  • Add to Cart