COMH421176DF Combi-Blocks, Inc.

[2-(2-Chlorophenyl)-1,3-thiazol-4-yl]acetic acid hydrochloride

  • MDL number MFCD19442563
  • Cas Number : 1984117-83-0
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7543
Availability details for
Quantity
  • Availability
  • Add to Cart