COMH421176E2 Combi-Blocks, Inc.

2-(3,3-Dibromo-1-methyl-2-oxopropyl)-1h-isoindole-1,3(2h)-dione

  • MDL number MFCD31583428
  • Cas Number : 32622-26-7
  • Molecular Weight : 375.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4,4-dibromo-3-oxobutan-2-yl)isoindoline-1,3-dione
Inchi Code 1S/C12H9Br2NO3/c1-6(9(16)10(13)14)15-11(17)7-4-2-3-5-8(7)12(15)18/h2-6,10H,1H3
Inchi Key IDPAFQPINBFLQW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7547
Availability details for
Quantity
  • Availability
  • Add to Cart