COMH421176E4 Combi-Blocks, Inc.

2-Amino-5-(3-nitrophenyl)furan-3,4-dicarbonitrile

  • MDL number MFCD31557133
  • Cas Number : 213836-68-1
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-(3-nitrophenyl)-3,4-furandicarbonitrile
Inchi Code 1S/C12H6N4O3/c13-5-9-10(6-14)12(15)19-11(9)7-2-1-3-8(4-7)16(17)18/h1-4H,15H2
Inchi Key MHJXVNVDXPLLRH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7552
Availability details for
Quantity
  • Availability
  • Add to Cart