COMH421176EA Combi-Blocks, Inc.

3-Morpholin-4-yl-5-phenylisoxazole-4-carboxylic acid

  • MDL number MFCD31583405
  • Cas Number : 2270905-18-3
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7587
Availability details for
Quantity
  • Availability
  • Add to Cart