COMH421176F0 Combi-Blocks, Inc.

4-(2,5-Dimethyl-1h-pyrrol-1-yl)butanoic acid

  • MDL number MFCD01688656
  • Cas Number : 110525-48-9
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7609
Availability details for
Quantity
  • Availability
  • Add to Cart