COMH421176FA Combi-Blocks, Inc.

2-(5-Methyl-1h-tetrazol-1-yl)benzoic acid

  • MDL number MFCD09035873
  • Cas Number : 72470-51-0
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7661
Availability details for
Quantity
  • Availability
  • Add to Cart