COMH42117700 Combi-Blocks, Inc.

Aniline blue

  • MDL number MFCD00036130
  • Cas Number : 28631-66-5
  • Molecular Weight : 737.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sodium 2-amino-3-methyl-5-((Z)-((Z)-4-((4-sulfonatophenyl)imino)cyclohexa-2,5-dien-1-ylidene)(4-((4-sulfophenyl)amino)phenyl)methyl)benzenesulfonate
Inchi Code 1S/C32H27N3O9S3.2Na/c1-20-18-23(19-30(32(20)33)47(42,43)44)31(21-2-6-24(7-3-21)34-26-10-14-28(15-11-26)45(36,37)38)22-4-8-25(9-5-22)35-27-12-16-29(17-13-27)46(39,40)41;;/h2-19,34H,33H2,1H3,(H,36,37,38)(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b31-22-,35-25-;;
Inchi Key XOSXWYQMOYSSKB-AETMCFOMSA-L

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7694
Availability details for
Quantity
  • Availability
  • Add to Cart