COMH42117704 Combi-Blocks, Inc.

3,4-Dibromo-mal-peg8-acid

  • MDL number MFCD30527391
  • Molecular Weight : 679.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C23H37Br2NO12/c24-20-21(25)23(30)26(22(20)29)2-4-32-6-8-34-10-12-36-14-16-38-18-17-37-15-13-35-11-9-33-7-5-31-3-1-19(27)28/h1-18H2,(H,27,28)
Inchi Key LDHUQDLPYALMHW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7718
Availability details for
Quantity
  • Availability
  • Add to Cart