COMH42117708 Combi-Blocks, Inc.

3-Tridecyl-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619044
  • Cas Number : 113737-17-0
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7729
Availability details for
Quantity
  • Availability
  • Add to Cart