COMH4211770A Combi-Blocks, Inc.

7-(2-Aminoethoxy)-3,4-dihydroquinolin-2(1h)-one hydrochloride

  • MDL number MFCD31583458
  • Cas Number : 2270908-52-4
  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7734
Availability details for
Quantity
  • Availability
  • Add to Cart