COMH42117714 Combi-Blocks, Inc.

(2-Pyridyldithio)-peg4-propargyl

  • MDL number MFCD31580154
  • Cas Number : 2170240-99-8
  • Molecular Weight : 357.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7794
Availability details for
Quantity
  • Availability
  • Add to Cart