COMH42117717 Combi-Blocks, Inc.

2,4-Dichloro-5-(piperidin-1-ylsulfonyl)benzoic acid

  • MDL number MFCD00760908
  • Cas Number : 53552-35-5
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7806
Availability details for
Quantity
  • Availability
  • Add to Cart