COMH4211771E Combi-Blocks, Inc.

Ethyl 4-methyl-2-[(3-methylphenyl)amino]pyrimidine-5-carboxylate

  • MDL number MFCD08592509
  • Cas Number : 908553-67-3
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7842
Availability details for
Quantity
  • Availability
  • Add to Cart