COMH42117729 Combi-Blocks, Inc.

2',6'-Dihydroxy-4'-methoxyacetophenone

  • MDL number MFCD00017518
  • Cas Number : 7507-89-3
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-dihydroxy-4-methoxyphenyl)ethanone
Inchi Code 1S/C9H10O4/c1-5(10)9-7(11)3-6(13-2)4-8(9)12/h3-4,11-12H,1-2H3
Inchi Key GKSGTWUNURZTKD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7905
Availability details for
Quantity
  • Availability
  • Add to Cart