COMH42117732 Combi-Blocks, Inc.

1-Hydroxy-2-butanone acetate

  • MDL number MFCD00027021
  • Cas Number : 1575-57-1
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7953
Availability details for
Quantity
  • Availability
  • Add to Cart