COMH42117735 Combi-Blocks, Inc.

M-Peg8-(ch2)12-phosphonic acid ethyl ester

  • MDL number MFCD30730359
  • Cas Number : 2112737-70-7
  • Molecular Weight : 644.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7972
Availability details for
Quantity
  • Availability
  • Add to Cart