COMH4211773D Combi-Blocks, Inc.

2-[4-(Pyrrolidin-1-ylsulfonyl)phenyl]ethanamine hydrochloride

  • MDL number MFCD23137024
  • Cas Number : 1224170-51-7
  • Molecular Weight : 290.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8029
Availability details for
Quantity
  • Availability
  • Add to Cart