COMH42117747 Combi-Blocks, Inc.

Methyl 2-(2-acetamidophenoxy)acetate

  • MDL number MFCD00525984
  • Cas Number : 452346-27-9
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8073
Availability details for
Quantity
  • Availability
  • Add to Cart