COMH4211774A Combi-Blocks, Inc.

2-((2-[1-(Methoxymethyl)-1h-1,2,4-triazol-5-yl]hydrazino)carbonyl)benzoic acid

  • MDL number MFCD31557138
  • Cas Number : 2199407-49-1
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8096
Availability details for
Quantity
  • Availability
  • Add to Cart