COMH4211774B Combi-Blocks, Inc.

Methyltetrazine-peg8-acid

  • MDL number MFCD31580146
  • Cas Number : 2183440-33-5
  • Molecular Weight : 612.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8098
Availability details for
Quantity
  • Availability
  • Add to Cart