COMH4211774C Combi-Blocks, Inc.

[3-Methoxy-4-(3-piperidin-1-ylpropoxy)phenyl]amine dihydrochloride

  • MDL number MFCD08582787
  • Cas Number : 1050556-41-6
  • Molecular Weight : 337.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8099
Availability details for
Quantity
  • Availability
  • Add to Cart