COMH42117754 Combi-Blocks, Inc.

(2-Pyridyldithio)-peg4-alcohol

  • MDL number MFCD31561123
  • Cas Number : 851961-99-4
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8152
Availability details for
Quantity
  • Availability
  • Add to Cart