COMH42117756 Combi-Blocks, Inc.

[2-(1H-Indol-1-yl)ethyl]amine sulfate

  • MDL number MFCD27756378
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8162
Availability details for
Quantity
  • Availability
  • Add to Cart