COMH42117758 Combi-Blocks, Inc.

3-(Diaminomethylene)furan-2,4(3h,5h)-dione

  • MDL number MFCD16992557
  • Cas Number : 1228552-29-1
  • Molecular Weight : 142.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8166
Availability details for
Quantity
  • Availability
  • Add to Cart