COMH4211775A Combi-Blocks, Inc.

(2-([(4-Fluorophenyl)sulfonyl]amino)-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD09047771
  • Cas Number : 926202-75-7
  • Molecular Weight : 316.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8172
Availability details for
Quantity
  • Availability
  • Add to Cart